Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AJ490336.1 | - | 5 - 6 | 1094 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Vitis vinifera Probe spans exons |