Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP3K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY461710.1 | - | 16 - 17 | 1974 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Vitis vinifera Probe spans exons |