Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY463369.1 | - | 1 | 544 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Vitis vinifera Both primers and probe map within a single exon |