Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | EF193002.3 | - | 1 - 2 | 123 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Vitis vinifera Probe spans exons |