Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | EU925561.1 | - | 15 - 16 | 1537 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Vitis vinifera Probe spans exons |