Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lrrc59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001205.1 | NP_001001205.1 | 3 - 4 | 266 | 70 | |
XM_018097691.1 | XP_017953180.1 | 2 - 3 | 947 | 70 | ||
GenBank mRNA | BC067310.1 | - | 3 - 4 | 266 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |