Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tram1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001210.2 | NP_001001210.1 | 2 - 3 | 277 | 76 | |
GenBank mRNA | BC067965.2 | - | 2 - 3 | 277 | 76 | |
CR760366.2 | - | 2 - 3 | 245 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |