Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: csnk1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001221.2 | NP_001001221.1 | 7 - 8 | 1080 | 80 | |
XM_012958779.2 | XP_012814233.1 | 8 - 9 | 1310 | 80 | ||
XM_012958780.2 | XP_012814234.1 | 8 - 9 | 1311 | 80 | ||
XM_012958782.2 | XP_012814236.1 | 8 - 9 | 1310 | 80 | ||
XM_012958784.2 | XP_012814238.1 | 7 - 8 | 1222 | 80 | ||
XM_018092013.1 | XP_017947502.1 | 8 - 9 | 1312 | 80 | ||
XM_018092014.1 | XP_017947503.1 | 7 - 8 | 1224 | 80 | ||
GenBank mRNA | BC067926.2 | - | 7 - 8 | 1080 | 80 | |
CT025205.2 | - | 7 - 8 | 1004 | 80 | ||
CT030065.2 | - | 7 - 8 | 1062 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |