Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fut1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004772.1 | NP_001004772.1 | 1 | 954 | 75 | |
XM_012966341.2 | XP_012821795.1 | 3 | 2506 | 75 | ||
XM_012966342.2 | XP_012821796.1 | 3 | 1645 | 75 | ||
XM_012966344.2 | XP_012821798.1 | 3 | 2351 | 75 | ||
XM_018095331.1 | XP_017950820.1 | 5 | 1481 | 75 | ||
XM_018095332.1 | XP_017950821.1 | 2 | 1531 | 75 | ||
GenBank mRNA | AJ781410.1 | - | 1 | 954 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Xenopus tropicalis Both primers and probe map within a single exon |