Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stoml2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004808.1 | NP_001004808.1 | 636 | 93 | ||
GenBank mRNA | BC074573.1 | - | 636 | 93 | ||
CR761170.2 | - | 628 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis |