Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004859.1 | NP_001004859.1 | 4 - 5 | 376 | 85 | |
XM_012968198.2 | XP_012823652.1 | 4 - 5 | 441 | 85 | ||
GenBank mRNA | BC074687.1 | - | 4 - 5 | 376 | 85 | |
CR855591.2 | - | 4 - 5 | 354 | 85 | ||
CR926234.1 | - | 4 - 5 | 358 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |