Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mpv17l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004930.1 | NP_001004930.1 | 4 - 5 | 545 | 97 | |
XM_012959523.2 | XP_012814977.1 | 3 - 4 | 317 | 97 | ||
GenBank mRNA | BC075397.1 | - | 4 - 5 | 545 | 97 | |
CR761343.2 | - | 4 - 5 | 536 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |