Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gapdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004949.2 | NP_001004949.1 | 8 - 9 | 777 | 75 | |
XM_012966386.2 | XP_012821840.1 | 9 - 10 | 991 | 75 | ||
GenBank mRNA | BC075438.1 | - | 8 - 9 | 778 | 75 | |
CR760856.2 | - | 8 - 9 | 793 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |