Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tdg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004982.1 | NP_001004982.1 | 9 - 10 | 1249 | 102 | |
XM_012958838.2 | XP_012814292.1 | 9 - 10 | 1324 | 102 | ||
XM_012958839.2 | XP_012814293.1 | 9 - 10 | 1200 | 102 | ||
GenBank mRNA | BC075533.1 | - | 9 - 10 | 1249 | 102 | |
CT030631.1 | - | 9 - 10 | 1242 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |