Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ankrd40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004991.1 | NP_001004991.1 | 4 - 5 | 847 | 90 | |
XM_012971643.2 | XP_012827097.1 | 3 - 4 | 1192 | 90 | ||
XM_012971644.2 | XP_012827098.1 | 4 - 5 | 995 | 90 | ||
XM_012971645.2 | XP_012827099.1 | 4 - 5 | 984 | 90 | ||
XM_012971646.2 | XP_012827100.1 | 4 - 5 | 963 | 90 | ||
GenBank mRNA | BC075561.1 | - | 4 - 5 | 847 | 90 | |
CR761248.1 | - | 3 - 4 | 999 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |