Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005010.1 | NP_001005010.1 | 3 - 4 | 477 | 93 | |
XM_012966413.2 | XP_012821867.1 | 3 - 4 | 510 | 93 | ||
GenBank mRNA | BC076663.1 | - | 3 - 4 | 477 | 93 | |
CR761011.2 | - | 3 - 4 | 474 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |