Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpl18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011030.2 | NP_001011030.1 | 5 - 6 | 510 | 75 | |
GenBank mRNA | BC082960.1 | - | 5 - 6 | 511 | 75 | |
CR760488.2 | - | 5 - 6 | 449 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |