Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203523.1 | NP_988854.1 | 8 - 9 | 1166 | 65 | |
GenBank mRNA | AY079192.1 | - | 8 - 9 | 1166 | 65 | |
CR926385.2 | - | 8 - 9 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Xenopus tropicalis Probe spans exons |