Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pnhd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203534.1 | NP_988865.1 | 1 - 2 | 96 | 66 | |
GenBank mRNA | AY333424.1 | - | 1 - 2 | 96 | 66 | |
BC170803.1 | - | 1 - 2 | 87 | 66 | ||
BC170805.1 | - | 1 - 2 | 87 | 66 | ||
CR926370.2 | - | 1 - 2 | 72 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |