Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203632.1 | NP_988963.1 | 6 - 7 | 789 | 96 | |
XM_012958113.2 | XP_012813567.1 | 6 - 7 | 860 | 96 | ||
GenBank mRNA | BC061436.1 | - | 6 - 7 | 789 | 96 | |
CR761464.2 | - | 5 - 6 | 771 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |