Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppid | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203653.1 | NP_988984.1 | 4 - 5 | 591 | 96 | |
GenBank mRNA | BC061335.1 | - | 4 - 5 | 591 | 96 | |
CT030612.1 | - | 4 - 5 | 532 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |