Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: etf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203704.1 | NP_989035.1 | 5 - 6 | 686 | 79 | |
GenBank mRNA | BC061387.1 | - | 5 - 6 | 686 | 79 | |
CR762244.2 | - | 4 - 5 | 543 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |