Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: adss | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203716.1 | NP_989047.1 | 12 - 13 | 1459 | 98 | |
GenBank mRNA | BC061354.1 | - | 12 - 13 | 1459 | 98 | |
CR761467.2 | - | 12 - 13 | 1326 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |