Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fkbp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203721.1 | NP_989052.1 | 5 - 6 | 646 | 98 | |
XM_018091140.1 | XP_017946629.1 | 5 - 6 | 671 | 98 | ||
GenBank mRNA | BC061340.1 | - | 5 - 6 | 646 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |