Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sfrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203731.1 | NP_989062.1 | 1 - 2 | 700 | 76 | |
GenBank mRNA | BC061311.1 | - | 1 - 2 | 700 | 76 | |
CR848123.2 | - | 1 - 2 | 753 | 76 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |