Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpsa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203737.1 | NP_989068.1 | 6 - 7 | 840 | 90 | |
GenBank mRNA | BC061298.1 | - | 6 - 7 | 840 | 90 | |
BC168799.1 | - | 5 - 6 | 816 | 90 | ||
CR942700.2 | - | 6 - 7 | 830 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |