Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atp1b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203739.1 | NP_989070.1 | 1 - 2 | 213 | 98 | |
GenBank mRNA | BC061283.1 | - | 1 - 2 | 229 | 98 | |
CR760087.2 | - | 1 - 2 | 196 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |