Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ak8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203773.1 | NP_989104.1 | 2 - 3 | 177 | 85 | |
GenBank mRNA | BC062516.1 | - | 2 - 3 | 177 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |