Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mapre1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203784.1 | NP_989115.1 | 4 - 5 | 612 | 87 | |
XM_012971600.2 | XP_012827054.1 | 6 - 7 | 841 | 87 | ||
XM_012971601.2 | XP_012827055.1 | 6 - 7 | 799 | 87 | ||
XM_012971602.2 | XP_012827056.1 | 6 - 7 | 817 | 87 | ||
XM_012971603.2 | XP_012827057.1 | 6 - 7 | 775 | 87 | ||
XM_012971604.2 | XP_012827058.1 | 5 - 6 | 758 | 87 | ||
XM_018090098.1 | XP_017945587.1 | 6 - 7 | 759 | 87 | ||
XM_018090099.1 | XP_017945588.1 | 5 - 6 | 627 | 87 | ||
GenBank mRNA | BC061382.1 | - | 4 - 5 | 612 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |