Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sf3b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203785.2 | NP_989116.1 | 5 - 6 | 1030 | 87 | |
GenBank mRNA | BC061357.1 | - | 5 - 6 | 1031 | 87 | |
CR760335.2 | - | 5 - 6 | 1001 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |