Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vgll2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203847.1 | NP_989178.1 | 3 - 4 | 624 | 69 | |
XM_012962842.2 | XP_012818296.1 | 2 - 3 | 769 | 69 | ||
XM_012962843.2 | XP_012818297.1 | 2 - 3 | 774 | 69 | ||
XM_012962844.2 | XP_012818298.1 | 2 - 3 | 563 | 69 | ||
XM_012962845.2 | XP_012818299.1 | 2 - 3 | 506 | 69 | ||
GenBank mRNA | BC063357.1 | - | 3 - 4 | 624 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |