Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: otx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203885.1 | NP_989216.1 | 1 - 2 | 210 | 82 | |
XM_012962849.2 | XP_012818303.1 | 2 - 3 | 335 | 82 | ||
GenBank mRNA | BC063197.1 | - | 1 - 2 | 210 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |