Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cfap45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203932.1 | NP_989263.1 | 9 - 10 | 1159 | 73 | |
XM_012969824.2 | XP_012825278.1 | 9 - 10 | 1456 | 73 | ||
GenBank mRNA | BC064248.1 | - | 9 - 10 | 1159 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |