Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lrr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203947.1 | NP_989278.1 | 1 - 2 | 255 | 86 | |
GenBank mRNA | BC064158.1 | - | 1 - 2 | 255 | 86 | |
CR761436.2 | - | 1 - 2 | 204 | 86 | ||
CU024931.1 | - | 1 - 2 | 211 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |