Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: copz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203948.1 | NP_989279.1 | 8 - 9 | 513 | 76 | |
XM_012956670.2 | XP_012812124.1 | 9 - 10 | 620 | 76 | ||
XM_012956671.2 | XP_012812125.1 | 8 - 9 | 581 | 76 | ||
XM_012956672.2 | XP_012812126.1 | 9 - 10 | 560 | 76 | ||
GenBank mRNA | BC064149.1 | - | 8 - 9 | 513 | 76 | |
CR760836.2 | - | 8 - 9 | 497 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |