Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bzw1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203950.1 | NP_989281.1 | 5 - 6 | 518 | 90 | |
GenBank mRNA | BC064244.1 | - | 5 - 6 | 518 | 90 | |
BC084993.1 | - | 4 - 5 | 429 | 90 | ||
CR761210.2 | - | 5 - 6 | 458 | 90 |
Gene Symbol: tent4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC168133.1 | - | 2494 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |