Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: actr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204037.1 | NP_989368.1 | 4 - 5 | 354 | 79 | |
GenBank mRNA | BC064225.1 | - | 4 - 5 | 354 | 79 | |
BC075462.1 | - | 4 - 5 | 355 | 79 | ||
CT485777.1 | - | 4 - 5 | 354 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |