Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213679.2 | NP_998844.1 | 6 - 7 | 1318 | 83 | |
GenBank mRNA | BC067963.2 | - | 6 - 7 | 1318 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |