Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rnf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213707.2 | NP_998872.1 | 3 - 4 | 553 | 88 | |
GenBank mRNA | BC067955.1 | - | 3 - 4 | 553 | 88 | |
BC161443.1 | - | 3 - 4 | 365 | 88 | ||
BC170688.1 | - | 3 - 4 | 523 | 88 | ||
BC171333.1 | - | 3 - 4 | 523 | 88 | ||
CU469377.1 | - | 3 - 4 | 349 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |