Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mrto4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005116.2 | NP_001005116.1 | 292 | 74 | ||
GenBank mRNA | BC077055.1 | - | 294 | 74 | ||
CR760103.2 | - | 283 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis |