Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005694.1 | NP_001005694.1 | 4 - 5 | 568 | 69 | |
GenBank mRNA | BC075257.1 | - | 4 - 5 | 568 | 69 | |
CR761857.2 | - | 4 - 5 | 618 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |