Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: krt70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006686.1 | NP_001006687.1 | 4 - 5 | 801 | 75 | |
GenBank mRNA | BC074628.1 | - | 4 - 5 | 801 | 75 | |
CR760223.2 | - | 4 - 5 | 788 | 75 | ||
CT485755.1 | - | 4 - 5 | 791 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |