Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pgpep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006751.1 | NP_001006752.1 | 4 - 5 | 882 | 94 | |
GenBank mRNA | BC075524.1 | - | 4 - 5 | 882 | 94 | |
CT030462.1 | - | 4 - 5 | 500 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |