Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: syngr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006784.1 | NP_001006785.1 | 2 - 3 | 432 | 76 | |
GenBank mRNA | BC075595.1 | - | 2 - 3 | 432 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |