Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006807.1 | NP_001006808.1 | 7 - 8 | 1000 | 81 | |
GenBank mRNA | BC076683.1 | - | 7 - 8 | 1000 | 81 | |
CR761150.2 | - | 7 - 8 | 1022 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |