Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: edar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006819.1 | NP_001006820.1 | 7 - 8 | 850 | 101 | |
GenBank mRNA | BC076874.1 | - | 7 - 8 | 850 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |