Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: calcoco2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006827.1 | NP_001006828.1 | 6 - 7 | 834 | 97 | |
GenBank mRNA | BC076895.1 | - | 6 - 7 | 834 | 97 | |
CR926335.2 | - | 6 - 7 | 877 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |