Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srd5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006840.1 | NP_001006841.1 | 3 - 4 | 830 | 104 | |
GenBank mRNA | BC076920.1 | - | 3 - 4 | 830 | 104 | |
CR761038.2 | - | 3 - 4 | 614 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |