Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdcd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006849.1 | NP_001006850.1 | 7 - 8 | 755 | 125 | |
GenBank mRNA | BC076936.1 | - | 7 - 8 | 755 | 125 | |
CR848219.2 | - | 706 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |