Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: phf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006860.1 | NP_001006861.1 | 2 - 3 | 588 | 101 | |
XM_012966441.2 | XP_012821895.1 | 2 - 3 | 776 | 101 | ||
XM_012966442.2 | XP_012821896.1 | 2 - 3 | 241 | 101 | ||
GenBank mRNA | BC076968.1 | - | 2 - 3 | 588 | 101 | |
CT030461.1 | - | 2 - 3 | 569 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |